Learn More
Sigma Aldrich Fine Chemicals Biosciences Saporin from Saponaria off5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S98965MG
Encompass
Saporin from Saponaria officinalis seeds has an N-terminal domain which is -stranded and a C-terminal domain which is helical. It is made up of 253 amino acids and has a molecular weight of 28 621Da.
Catalog No. 50-290-5370