Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sapropterin (dihydrochloride) | 69056-38-8 | 99.8% | C9H17Cl2N5O3 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYA0124A

Encompass_Preferred

Sapropterin (dihydrochloride) is an orally active phenylalanine hydroxylase (PAH) cofactor, effective in reducing blood phenylalanine concentrations. It is used in the study of phenylketonuria (PKU) and can also aggravate experimental autoimmune encephalomyelitis (EAE). This product appears as a white to off-white solid.

  • CAS number: 69056-38-8
  • Chemical formula: C9H17Cl2N5O3
  • Purity (LCMS): 99.8%
  • Target: Endogenous Metabolite
  • Pathway: Metabolic Enzyme/Protease
  • Storage at 4°C, stored under nitrogen, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month.
  • Soluble in H2O and DMSO at 100 mg/mL.

Catalog No. 50-003-59056


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.