Learn More
Medchemexpress LLC Sapropterin (dihydrochloride) | 69056-38-8 | 99.8% | C9H17Cl2N5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA0124A5MG
Encompass_Preferred
Sapropterin dihydrochloride is an orally active phenylalanine hydroxylase (PAH) cofactor that effectively reduces blood phenylalanine concentrations. It is utilized in the study of phenylketonuria (PKU) and has been shown to aggravate experimental autoimmune encephalomyelitis (EAE).
  • Orally active phenylalanine hydroxylase (PAH) cofactor
  • Reduces blood phenylalanine concentrations
  • Used in phenylketonuria (PKU) research
  • Aggravates experimental autoimmune encephalomyelitis (EAE)
  • Enhances residual phenylalanine hydroxylase activity
Catalog No. 50-004-32808