Learn More
Medchemexpress LLC Sapropterin dihydrochloride | 69056-38-8 | 99.8% | C9H17Cl2N5O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA0124A50MG
Encompass_Preferred
Sapropterin ((6R)-BH4; (6R)-Tetrahydro-L-biopterin) dihydrochloride is an orally active phenylalanine hydroxylase (PAH) cofactor. It is effective in reducing blood phenylalanine concentrations and can be used in the study of phenylketonuria (PKU). Additionally, Sapropterin dihydrochloride can aggravate experimental autoimmune encephalomyelitis (EAE).
  • Orally active phenylalanine hydroxylase (PAH) cofactor.
  • Effective in reducing blood phenylalanine concentrations.
  • Used in the study of phenylketonuria (PKU).
  • Can aggravate experimental autoimmune encephalomyelitis (EAE).
Catalog No. 50-004-32221