Learn More
Medchemexpress LLC Sapropterin dihydrochloride | 69056-38-8 | 99.9% | C9H17Cl2N5O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYA0124AR1MG
Encompass_Preferred
Sapropterin dihydrochloride is an analytical standard intended for research and analytical applications. This compound is an orally active phenylalanine hydroxylase (PAH) cofactor that effectively reduces blood phenylalanine concentrations. It can be used in the study of phenylketonuria (PKU) and may also aggravate experimental autoimmune encephalomyelitis (EAE).
  • Analytical standard for research and analytical applications.
  • Orally active phenylalanine hydroxylase (PAH) cofactor.
  • Effectively reduces blood phenylalanine concentrations.
  • Used in the study of phenylketonuria (PKU).
  • May aggravate experimental autoimmune encephalomyelitis (EAE).
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-004-32915