Learn More
Medchemexpress LLC Saquinavir mesylate | 149845-06-7 | 99.8% | 766.95 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1700310MM/1ML
Encompass_Preferred
Saquinavir mesylate is an orally active HIV protease inhibitor used in AIDS research. It also exhibits anti-inflammatory activity and can induce apoptosis of human red blood cells.
  • Orally active HIV protease inhibitor
  • Used in AIDS research
  • Exhibits anti-inflammatory activity
  • Can induce apoptosis of human red blood cells
Catalog No. 50-003-90708