Learn More
Medchemexpress LLC (Sar1)-Angiotensin II | 51833-69-3 | 99.8% | 1002.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP31385MG
Encompass_Preferred
(Sar1)-Angiotensin II, an analogue of Angiotensin II, is a specific agonist of angiotensin AT1 receptor. It binds to brain membrane-rich particles with a Kd of 2.7 nM and can stimulate protein synthesis and cell growth in embryonic chick myocytes.
  • Analogue of Angiotensin II
  • Specific agonist of angiotensin AT1 receptor
  • Binds to brain membrane-rich particles with a Kd of 2.7 nM
  • Can stimulate protein synthesis and cell growth in embryonic chick myocytes
Catalog No. 50-003-96630