Learn More
Apexbio Technology LLC SAR191801 1234708-04-3 5mg
SDP
Supplier:  Apexbio Technology LLC B58405
Encompass_Preferred
SAR191801 (CAS 1234708-04-3) is a selective inhibitor of human hematopoietic prostaglandin D synthase (hPGDS) By targeting and suppressing hPGDS enzymatic activity SAR191801 effectively reduces the synthesis of prostaglandin D (PGD ) In fluorescence polarization and enzyme immunoassay experiments it demonstrated an IC value of 12 nM for hPGDS inhibition SAR191801 serves as a valuable tool for studying physiological processes and pathologies mediated by PGD including allergic reactions inflammatory responses and sleep-wake regulation mechanisms
Catalog No. 50-000-90553