Learn More
Apexbio Technology LLC SAR245409 (XL765) 1349796-36-6 100mg
SDP
Supplier:  Apexbio Technology LLC A5634100
Encompass_Preferred
SAR245409 (XL765 CAS 1349796-36-6) is a selective dual inhibitor of phosphatidylinositol-3-kinase (PI3K) and mammalian target of rapamycin (mTOR) with high potency against PI3K (IC50 9 nM) It suppresses the PI3K/Akt/mTOR signaling cascade by preventing formation of phosphatidylinositol-3 4 5-triphosphate (PIP3) at the cell membrane and downstream phosphorylation of AKT p70S6 kinase and S6 SAR245409 demonstrates antitumor activity in cancer models exhibiting genetic alterations in PI3K pathways enhancing growth suppression and apoptosis It is commonly employed in preclinical studies evaluating PI3K/mTOR-targeted approaches in cancer therapy
Catalog No. 50-000-61468