Learn More
Medchemexpress LLC HY-12481A 2mg Medchemexpress, SAR405 R enantiomer CAS:1946010-79-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12481A2MG
Encompass_Preferred
Medchemexpress, HY-12481A 2mg SAR405 R enantiomer CAS:1946010-79-2 SAR405 R enantiomer is the less active enantiomer of SAR405. SAR405 is a PIK3C3/Vps34 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2598