Learn More
ABclonal Technology SARA Rabbit mAb
SDP
Supplier:  ABclonal Technology A20900100UL
Encompass_Preferred
This gene encodes a double zinc finger motif-containing protein that participates in the transforming growth factor-beta (TGFB) signalling pathway The encoded protein interacts directly with SMAD2 and SMAD3 and recruits SMAD2 to the TGFB receptor There are multiple pseudogenes for this gene on chromosomes 2 15 and X Alternative splicing results in multiple transcript variants
Catalog No. 50-000-07923