Learn More
Cayman Chemical Saracatnb 5mg
SDP
Supplier:  Cayman Chemical 114975 MG
Encompass_Preferred
A dual inhibitor of c-Src and Abl (IC50 = 2.7 and 30 nM, respectively); less effectively inhibits other kinases; blocks cell motility, migration, adhesion, invasion, proliferation, differentiation, and survival; reduces osteoclast bone resorption
Catalog No. 50-295-4575