Learn More
Medchemexpress LLC Sarafloxacin hydrochloride | 91296-87-6 | 98.1% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0343A1G
Encompass_Preferred
Sarafloxacin hydrochloride is a quinolone antibiotic agent, specifically a fluoroquinolone, registered for use in treating poultry diseases. It has demonstrated efficacy in field trials for treating channel catfish infected with E. ictaluri.
  • Quinolone antibiotic agent.
  • Fluoroquinolone antibiotic.
  • Efficacious in treating channel catfish infected with E. ictaluri.
Catalog No. 50-004-06211