Learn More
Medchemexpress LLC Sarafloxacin hydrochloride | 91296-87-6 | 98.13% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0343A5G
Encompass_Preferred
Sarafloxacin hydrochloride is a quinolone antibiotic agent, effective against poultry diseases and *Edwardsiella ictaluri* infections in channel catfish. Its inhibitory levels vary by bacterial strain, with *Escherichia coli* isolates showing higher sensitivity in broth culture compared to simulated gut models, suggesting potential absorption to organic matter. In field trials, administration to catfish infected with *E. ictaluri* significantly improved survival rates.
  • Quinolone antibiotic agent
  • Effective against poultry diseases
  • Successfully treated *Edwardsiella ictaluri* infections in channel catfish
  • Demonstrated varying inhibitory levels for tested bacteria
  • Improved survival rates in field trials for infected catfish
Catalog No. 50-004-06279