Learn More
Medchemexpress LLC Sarafloxacin hydrochloride | 91296-87-6 | MFCD03427297 | 98.1% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB0343A10G
Encompass_Preferred
Sarafloxacin hydrochloride (A-56620 hydrochloride) is a quinolone antibiotic agent. It is a fluoroquinolone antibiotic registered for use against poultry diseases. It has been shown to be effective in treating E. ictaluri infections in channel catfish in vivo. This product is for research use only.
  • Quinolone antibiotic agent
  • Fluoroquinolone antibiotic for poultry diseases
  • Efficacious in treating E. ictaluri infections in channel catfish
  • Demonstrates mineralization in loam, silt loam, and sandy loam soils
  • Inhibitory level for tested bacteria is strain dependent
Catalog No. 50-004-07255