Learn More
Selleck Chemical LLC Saralasin Acetate P1150-5MG
SDP
Supplier:  Selleck Chemical LLC P11505MG
Encompass_Preferred
Saralasin Acetate an octapeptide analog of angiotensin II is a highly specific competitive inhibitor of angiotensin II that is used in the diagnosis of hypertension
Catalog No. 50-000-43626