Learn More
Medchemexpress LLC Sarcosine-15N | 99.9% | 90.09 g/mol | C3H715NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101037S5MG
Encompass_Preferred
Sarcosine-15N is the 15N-labeled isotopologue of sarcosine (N-methylglycine), provided as a high-purity analytical standard for metabolomics, isotopic tracing, and quantitative mass spectrometry. The labeled compound enables accurate internal standardization and tracer studies while retaining the chemical behavior of the native analyte.
  • High purity suitable for analytical applications.
  • Isotopically enriched 15N for tracer and metabolic studies.
  • Compatible with mass spectrometry and NMR as an internal standard.
  • Supplied in a small quantity appropriate for method development and reference use.
  • Stable solid form with standard storage recommendations for labeled amino acids.
Catalog No. 50-002-13566