Learn More
Medchemexpress LLC Sarcosine-d3 (N-methylglycine-d3) | 118685-91-9 | MFCD01075434 | 99.1% | 92.11 g·mol⁻¹ | C3H4D3NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101037S110MG
Encompass_Preferred
Sarcosine-d3 (N-methylglycine-d3) is a deuterium-labeled amino acid provided as a high-purity solid for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is supplied in small milligram quantities for research use with recommended storage guidance for powder and solutions.
  • High isotopic purity for accurate mass-spectrometry quantitation.
  • 99.05% purity as supplied.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Solid, colorless to off-white powder with high water solubility (≈125 mg/mL).
  • Stable as a powder at -20°C; solution stability depends on temperature.
  • Offered in small mg sizes appropriate for analytical workflows.
Catalog No. 50-002-12023