Learn More
Medchemexpress LLC Sarcosine-d3 (N-methylglycine-d3) | 118685-91-9 | MFCD01075434 | 99.1% | 92.11 g/mol | C3H4D3NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101037S1100MG
Encompass_Preferred
Sarcosine-d3 (N-methylglycine-d3) is a deuterium-labeled analog of sarcosine supplied as a colorless to off-white solid for research use. It serves as a tracer and quantitative internal standard for NMR, GC-MS, and LC-MS analyses, and is commonly used in studies of glycine transport and NMDA receptor modulation (CAS 118685-91-9).
  • Deuterium-labeled (d3) for accurate internal standardization.
  • High chemical purity (≈99.1%).
  • Water soluble (≈125 mg/mL); may require ultrasonic agitation.
  • Stable as a powder at -20°C; in solution, store at -80°C for extended periods.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS quantitation.
Catalog No. 50-002-12822