Learn More
Selleck Chemical LLC Sardomozide dihydrochloride
SDP
Supplier:  Selleck Chemical LLC E16941G
Encompass_Preferred
Sardomozide dihydrochloride (CGP 48664A) is a potent inhibitor of S-adenosylmethionine decarboxylase (SAMDC) with an IC50 of 5 nM It is also a potent inhibitor of polyamine oxidase and exhibits with broad spectrum antiproliferative and antitumor activity
Catalog No. 50-313-7174