Learn More
Medchemexpress LLC Sarecycline hydrochloride | 1035979-44-2 | 99.8% | 523.96 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13858A10MM/1ML
Encompass_Preferred
Sarecycline hydrochloride is an orally active, narrow-spectrum tetracycline derivative antibiotic with anti-inflammatory properties. It is known to inhibit Gram-positive bacteria and several types of *keratobacterium acnes*. Its mechanism of action involves interfering with tRNA accommodation and tethering mRNA to the 70S ribosome. This compound is used in research to study moderate to severe acne.
  • Orally active
  • Narrow-spectrum antibiotic
  • Anti-inflammatory activity
  • Mechanism of action involves interfering with tRNA accommodation and tethering mRNA to the 70S ribosome
  • Used for studying moderate to severe acne
Catalog No. 50-003-50414