Learn More
TARGETMOL CHEMICALS INC Sarizotan 2HCl 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T70044L1MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Sarizotan 2HCl is an hERG channel inhibitor and 5-hydroxytryptamine 5-HT1A receptor agonist with potential antidepressant effects for the study of Parkinsonian movement disorders. Purity 98.83%
Catalog No. 50-287-8102