Learn More
Medchemexpress LLC Sarpogrelate hydrochloride | 135159-51-2 | MFCD00887582 | 465.97 | C24H32ClNO6 | 5 MG

Supplier: Medchemexpress LLC HY105645MG
Sarpogrelate hydrochloride is the hydrochloride salt of a selective 5-HT2 receptor antagonist used in research to study vascular disease and thrombosis. It is characterized by reported pKi values for 5-HT2A, 5-HT2B, and 5-HT2C receptors and includes solubility and storage guidance for laboratory handling.
- Selective 5-HT2 receptor antagonist for vascular and thrombosis research.
- Reported pKi values: 5-HT2A = 8.52; 5-HT2B = 6.57; 5-HT2C = 7.43.
- Molecular formula C24H32ClNO6 and molecular weight 465.97.
- Soluble in DMSO and water; formulation options provided for in vivo use.
- Storage recommendations include 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.