Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike RBD Recombinant Protein, Endotoxin-Free
Supplier:  Sino Biological 40592VNAHUE1
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (RBD) (YP_009724390.1) (Arg319-Phe541) was expressed.
Catalog No. 50-000-84052