Learn More
Medchemexpress LLC SARS-CoV-2-IN-7 | 2570461-66-2 | 98.8% | C14H8F3NOSe | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141841100MG
Encompass_Preferred
SARS-CoV-2-IN-7 is an inhibitor that effectively inhibits viral replication with an IC50 of 844 nM in SARS-CoV-2-infected Vero E6 cells. It is intended for research use only and not sold to patients. The compound has a molecular weight of 342.17 and appears as a light yellow to orange solid.
  • Inhibits SARS-CoV-2 viral replication in Vero E6 cells (IC50 of 844 nM).
  • Exhibits cytotoxicity against human HaCaT cells (IC50 > 32 μM).
  • Highly soluble in DMSO (250 mg/mL) and forms clear solution with 10% DMSO and 90% corn oil (≥ 2.08 mg/mL) for in vivo experiments.
  • Powder storage: -20°C for 3 years or 4°C for 2 years.
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month.
  • Light yellow to orange solid.
Catalog No. 50-003-55262