Learn More
Medchemexpress LLC SARS-CoV-2-IN-86 | 368866-07-3 | MFCD02683117 | 99.4% | 466.53 | C26H30N2O6 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0244851G
Encompass_Preferred
SARS-CoV-2-IN-86 is a small-molecule research compound that inhibits SARS-CoV-2 methyltransferases (nsp14 and nsp16) and is reported as an andrographolide derivative. It is supplied as a solid or as a 10 mM solution in DMSO for in vitro antiviral and enzymatic studies. Batch-specific certificates of analysis and safety documentation are available from the supplier.
  • Inhibits SARS-CoV-2 methyltransferases (nsp14 and nsp16).
  • Andrographolide derivative for antiviral research.
  • Available as solid and 10 mM solution in DMSO.
  • High purity (≈99.4%) per supplier documentation.
  • Suitable for in vitro enzymatic and antiviral assays.
  • Powder stable at -20°C; solvent storage guidelines available.
Catalog No. 50-003-01244