Learn More
Sino Biological SARS-CoV-2 JN.1 (Omicron) Spike S1+S2 trimer Protein (ECD, His Tag)(MALS-verified)
Supplier:  Sino Biological 40589V08H59100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (JN.1) Spike S1+S2 (YP_009724390.1, with mutations ins16MPLF, T19I, R21T, L24del, P25del, P26del, A27S, S50L, H69del, V70del, V127F, G142D, Y144del, F157S, R158G, N211del, L212I, V213G, L216F, H245N, A264D, I332V, G339H, K356T, S371F, S373P, S375F, T376A, R403K, D405N, R408S, K417N, N440K, V445H, G446S, N450D, L452W, L455S, N460K, S477N, T478K, N481K, V483del, E484K, F486P, Q498R, N501Y, Y505H, E554K, A570V, D614G, P621S, H655Y, N679K, P681R, N764K, D796Y, F817P, A892P, A899P, S939F, A942P, Q954H, N969K, K986P, V987P, P1143L and furin cleavage site mutants) expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant JN.1).
Catalog No. NC2887637