Learn More
Cayman Chemical SARS-CoV-2 Main Protease Inhi
SDP
Supplier:  Cayman Chemical 7019641 EA
Encompass_Preferred
SARS-CoV-2 Main Protease Inhibitor (GC376) has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Main Protease Inhibitor Screening Assay Kit (Item No. 701960).
Catalog No. 50-289-3974