Learn More
Genscript Corporation SARS-CoV-2 Neutralizing Antibo
Supplier:  Genscript Corporation A02088100
Encompass
The product is specific for SARS-CoV-2 Spike Protein S1 subunit and its RBD domain The product can recognize and neutralize Wild-Type SARS-CoV-2 and Variants of Concern (VOC) including Alpha Beta Delta and Omicron Reactivity with Gamma variant has not been tested
Catalog No. 50-000-58368