Learn More
Cayman Chemical SARS-CoV-2 nutralIzng ntIb 1ea
SDP
Supplier:  Cayman Chemical 5020801 EA
Encompass_Preferred
A set of SARS-CoV-2 neutralizing antibody human plasma controls containing negative control plasma, as well as positive control plasma spiked with low, mid, and high levels of recombinant human anti-SARS-CoV-2 neutralizing IgG antibody
Catalog No. 50-289-5230