Learn More
Cayman Chemical SARS-CoV-2 nutralIzng ntIb 1ea
SDP
Supplier:  Cayman Chemical 5022021 EA
Encompass_Preferred
SARS-CoV-2 Neutralizing Antibody (B.1.351) Positive Control has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 (B.1.351, Beta Variant) Neutralizing Antibody SimpleDetect ELISA Kit (Item No. 502200).
Catalog No. 50-289-5124