Learn More
Medchemexpress LLC SARS-CoV-2 S glycopr 2ug
SDP
Supplier:  Medchemexpress LLC HYP720352UG
Encompass_Preferred
Recombinant SARS-CoV-2 S glycoprotein (residues V16-R685) produced in HEK293 cells, supplied as a glycosylated, affinity-tagged protein for use in serology, antibody characterization, and assay development. The material is provided in small pack sizes and handled to preserve structural and antigenic features relevant to immunoassays.
  • Produced in HEK293 mammalian cells to preserve native glycosylation.
  • Includes N-terminal 10xHis and C-terminal Flag tags for detection and purification.
  • Covers residues V16-R685 of the spike protein.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~120 kDa.
  • Supplied in small pack sizes; store at -20°C and aliquot for long-term storage.
  • After reconstitution, stable at 4°C for one week; freeze at -20°C or -80°C for extended storage.
  • Suitable for serology, antibody binding assays, and assay development.
Catalog No. 50-001-96000