Learn More
Medchemexpress LLC SARS-CoV-2 S protein receptor-binding domain, biotinylated (Omicron B.1.1.529) | 96.0% | 50 UG

Supplier: Medchemexpress LLC HYP7444650UG
Recombinant biotinylated receptor-binding domain (RBD) of the SARS-CoV-2 spike protein (Omicron B.1.1.529). Produced in HEK293 cells and supplied as a filtered PBS solution, this enzymatically biotinylated RBD includes C-terminal His and Avi tags and is intended for binding assays, ELISA development, and antibody characterization.
- Expression system: HEK293 cells
- Biotinylation: site-specific Avi tag
- Formulation: 0.22 μm filtered PBS, pH 7.4
- Concentration: 0.54 mg/mL
- Purity: 96.0% by reducing SDS-PAGE
- Endotoxin: <1 EU/μg (LAL)
- Storage: store at -80°C; avoid repeated freeze-thaw
- Applications: ACE2 binding assays, ELISA, antibody characterization
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.