Learn More
ABclonal Technology SARS-CoV-2 Spike RBD Mouse mAb
SDP
Supplier:  ABclonal Technology A2377120UL
Encompass_Preferred
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is an enveloped positive-sense single-stranded RNA virus that causes COVID-19 The virus contains RNA and structural proteins essential for host cell invasion Inside the host the RNA encodes structural proteins that form virus particles nonstructural proteins for virus assembly replication and host control and accessory proteins with unclear functions Structural proteins include the envelope (E) spike (S) membrane (M) and nucleocapsid (N) proteins The spike protein which gives the virus its crown-like appearance mediates attachment and entry into host cells It is a key target for vaccine development antibody therapies and diagnostic tests
Catalog No. 50-000-04879