Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD-AVI & His Recombinant Protein, Biotinylated
Supplier:  Sino Biological 40589V27BB20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S1+S2 ECD) (YP_009724390.1) (Met1-Pro1213) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5542