Learn More
ACROBiosystems SARS-CoV-2 Spike S2 protein, His Tag (BA.2/Omicron) (S2N-C52Hh) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (N764K, D796Y, Q954H, N969K, F817P, A892P, A899P, A942P, K986P, V987P)). T
Supplier:  ACROBiosystems S2NC52HH1MG
Encompass_Preferred
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Catalog No. 50-225-3610