Learn More
Medchemexpress LLC SARS-CoV S HEK293 50ug

Supplier: Medchemexpress LLC HYP7204550UG
Recombinant SARS-CoV spike (S) protein produced in human HEK293 cells and provided with affinity tags for detection or purification. The product is supplied at >95% purity and is intended for research use in antibody development, receptor binding assays, ELISA, and other immunoassay applications. Available in small-scale sizes, including a 50 μg format.
- Expressed in HEK293 human cells.
- Contains N-terminal 10xHis and C-terminal Myc tags.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Approximate molecular weight of ~37 kDa.
- Supplied in sizes suitable for assay development and small-scale research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.