Learn More
Selleck Chemical LLC Sarsasapogenin S3607-5mg
SDP
Supplier:  Selleck Chemical LLC S36075MG
Encompass_Preferred
Sarsasapogenin (SAR Parigenin) is a steroidal sapogenin It can provoke the generation of reactive oxygen species and activate unfolded protein response (UPR) signaling pathways SAR potently inhibits NF- B and MAPK activation as well as IRAK1 TAK1 and I B phosphorylation in LPS-stimulated macrophages
Catalog No. 50-310-8918