Learn More
Sino Biological SARS-CoV-2 B.1.1.529 (Omicron) Spike S1 NTD Protein (His Tag) 100ug
Supplier:  Sino Biological 40591V08H42100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1-NTD (YP_009724390.1, with mutations A67V, H69del, V70del, T95I, G142D, V143del, Y144del, Y145del, N211del, L212I) (Met1-Ser305) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.1.529) which emerged in the South Africa.
Catalog No. 50-222-1648