Learn More
Cayman Chemical SARSCoV2 SpIke GlycoproteIn
SDP
Supplier:  Cayman Chemical 30590100 UG
Encompass_Preferred
Source: Active recombinant C-terminal rabbit IgG1 Fc-tagged SARS-CoV-2 surface glycoprotein receptor binding domain expressed in HEK293 cells • Amino acids: 319-541 • MW: 50.3 kDa
Catalog No. 50-299-1985