Learn More
Cayman Chemical SARSCoV2 SpIkeAC2 InTRac 96
SDP
Supplier:  Cayman Chemical 50205096 SOLID
Encompass_Preferred
Cayman’s SARS-CoV-2 Spike-ACE2 Interaction Inhibitor Screening Assay Kit provides a robust and EAsy-to-use platform for identifying novel inhibitors of the SARS-CoV-2 spike and ACE2 interaction. The assay uses a recombinant rabbit Fc-tagged SARS-CoV-2 spike S1 RBD that binds to a plate precoated with a mouse anti-rabbit antibody. A recombinant His-tagged ACE2 protein binds the spike RBD and the complex is detected with an HRP-conjugated anti-His antibody, which is EAsily quantified by reading the absorbance at 450 nm. A control is included for competition of the SARS-CoV-2 spike RBD-ACE2 interaction.
Catalog No. 50-289-5284