Learn More
Cayman Chemical RacecadotrIl-d5 10mg
SDP
Supplier:  Cayman Chemical 3389310 MG
Encompass_Preferred
An internal standard for the quantification of racecadotril by GC- or LC-MS
Catalog No. 50-294-8755