Learn More
Sartorius SARTOPORE 2 CAPSULE PK4
Supplier:  Sartorius 5445307H7OOA/DEL
Encompass
Sartopore 2 is the industry standard of high-performance polyethersulfone (PES) liquid filters. a broad variety of different pore size combinations ensures highly economic and safe filtration performance. From single-layer high flow solutions to specific Mycoplasma retentive filters, Sartopore 2 perfectly adapts to your specific process needs.
Catalog No. NC9955756