Learn More
Cayman Chemical Sartrypyron D 100ug
SDP
Supplier:  Cayman Chemical 31676100 UG
Encompass_Preferred
A fungal metabolite; inhibits NFRD (IC50 = 1.7 µM in A. suum mitochondria) and NADH oxidase (IC50 = 3 µM in bovine heart mitochondria); active against B. subtilis, K. rhizophila, and M. smegmatis in disc assays
Catalog No. 50-298-8081