Learn More
Enterprise Technology Solutions Satco Products, Inc. Light Bulb,A15,40W f/Appliances,225Lumens,E26,WarmWhite
SDP
Supplier:  Enterprise Technology Solutions SDNS3720
Encompass_Preferred
A15-sized Appliance light bulb uses 40 watts and puts out 225 lumens. It has a medium base (E26) and is 120V/130V. Made to Commercial Quality Specifications, bulbs are ideal for use in ovens.
Catalog No. 50-007-89971