Learn More
Medchemexpress LLC Satigrel 100mg | 111753-73-2 | 100 MG

Supplier: Medchemexpress LLC HY114564100MG
Satigrel is a research-grade small-molecule inhibitor of prostaglandin-endoperoxide synthase 1 (PGHS1/COX-1) used to study platelet aggregation and prostaglandin signaling in vitro and preclinical assays.
- Selective PGHS1 (COX-1) inhibitor
- Reported IC50 for PGHS1: 0.081 μM
- Also inhibits PGHS2 and several phosphodiesterase isoforms
- Molecular formula C20H19NO4 and molecular weight 337.37 g/mol
- High purity (99.9%) suitable for research applications
- Available in multiple pack sizes, including 100 MG
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.