Learn More
Medchemexpress LLC Sauchinone | 177931-17-8 | 356.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN06135MG
Encompass_Preferred
Sauchinone is a diastereomeric lignan isolated from *Saururus chinensis* (Saururaceae). It inhibits LPS-inducible iNOS, TNF-α, and COX-2 expression by suppressing I-κBα phosphorylation and p65 nuclear translocation. Sauchinone demonstrates anti-inflammatory and antioxidant activities.
  • Anti-inflammatory activity
  • Antioxidant activity
Catalog No. 50-003-88376