Learn More
Medchemexpress LLC Sauchinone | 177931-17-8 | MFCD08702698 | 99.9% | 356.37 g/mol | C20H20O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN061310MG
Encompass_Preferred
Sauchinone is a diastereomeric lignan isolated from Saururus chinensis that inhibits NF-κB signaling and is used as a research reagent to study inflammatory pathways and related cellular responses. It is provided as a high-purity analytical standard suitable for in vitro assays.
  • Inhibits NF-κB signaling and suppresses LPS-inducible iNOS, TNF-α, and COX-2 expression.
  • Diastereomeric lignan; molecular formula C20H20O6; molecular weight 356.37 g/mol.
  • High purity appropriate for research applications.
  • Available in small analytical quantities, including 10 mg pack sizes.
  • For research use only; not for human consumption or diagnostic use.
Catalog No. 50-002-95938