Learn More
Enzo Life Sciences SAVIEW PLUS AP Reagent (150tests)
SDP
Supplier:  Enzo Life Sciences ENZACC1110150
Encompass_Preferred
SAVIEW PLUS AP Reagent is a ready-to-use streptavidin-based nanopolymer detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded (FFPE) tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the SAVIEW PLUS AP Reagent can be used for in situ hybridization detection of biotinylated DNA/RNA probes. Long Term Storage: +4°C. Assay Time: < 2 hours. Applications: IHC ISH (in situ hybridization).
Catalog No. 50-201-3359