Learn More
TARGETMOL CHEMICALS INC SAVOLITINIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ021010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Savolitinib (Volitinib) (AZD-6094) is an effective selective and orally bioavailable c-Met inhibitor (IC50s 5 nM/3 nM for c-Met/p-Met). purity: 98%
Catalog No. 50-257-6151